The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128217.2 | NP_001121689.1 | 10 - 11 | 1323 | 90 |
NM_033222.4 | NP_150091.2 | 10 - 11 | 1326 | 90 | |
GenBank mRNA | AF063020.1 | - | 10 - 11 | 1292 | 90 |
AF098483.1 | - | 9 - 10 | 1006 | 90 | |
AF432220.1 | - | 10 - 11 | 1274 | 90 | |
U94319.1 | - | 4 - 5 | 445 | 90 |