The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020778.4 | NP_065829.3 | 11 - 12 | 5272 | 58 |
XM_017022444.1 | XP_016877933.1 | 10 - 11 | 5270 | 58 | |
GenBank mRNA | AB037751.1 | - | 6 - 7 | 2221 | 58 |
AL832372.1 | - | 6 - 7 | 2509 | 58 | |
AL833276.1 | - | 6 - 7 | 2588 | 58 | |
AL833288.1 | - | 7 - 8 | 3954 | 58 | |
AY044449.1 | - | 11 - 12 | 5272 | 58 |