The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145860.1 | NP_665859.1 | 1 - 2 | 37 | 126 |
XM_017005645.1 | XP_016861134.1 | 1 - 2 | 885 | 126 | |
GenBank mRNA | AF022385.1 | - | 1 - 2 | 37 | 126 |