The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007217.3 | NP_009148.2 | 6 - 7 | 789 | 123 |
NM_145859.1 | NP_665858.1 | 6 - 7 | 638 | 123 | |
NM_145860.1 | NP_665859.1 | 5 - 6 | 544 | 123 | |
XM_005247086.4 | XP_005247143.1 | 6 - 7 | 1524 | 123 | |
XM_005247087.4 | XP_005247144.1 | 6 - 7 | 1529 | 123 | |
XM_005247088.3 | XP_005247145.1 | 5 - 6 | 620 | 123 | |
XM_006713485.3 | XP_006713548.1 | 6 - 7 | 684 | 123 | |
XM_011512368.2 | XP_011510670.1 | 7 - 8 | 1597 | 123 | |
XM_011512369.2 | XP_011510671.1 | 7 - 8 | 1602 | 123 | |
XM_017005644.1 | XP_016861133.1 | 6 - 7 | 679 | 123 | |
XM_017005645.1 | XP_016861134.1 | 6 - 7 | 1520 | 123 | |
GenBank mRNA | AF022385.1 | - | 5 - 6 | 544 | 123 |
AK291130.1 | - | 5 - 6 | 575 | 123 | |
BC002506.1 | - | 6 - 7 | 801 | 123 | |
BC016353.1 | - | 6 - 7 | 642 | 123 | |
CR457107.1 | - | 4 - 5 | 391 | 123 |