The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181706.4 | NP_859057.4 | 2 - 3 | 230 | 148 |
XM_017017216.1 | XP_016872705.1 | 2 - 3 | 215 | 148 | |
GenBank mRNA | AK123087.1 | - | 2 - 3 | 221 | 148 |
AK289665.1 | - | 2 - 3 | 218 | 148 | |
AL833128.1 | - | 2 - 3 | 220 | 148 | |
BC036571.1 | - | 2 - 3 | 205 | 148 | |
BC044931.2 | - | 2 - 3 | 185 | 148 | |
BC063804.1 | - | 2 - 3 | 173 | 148 | |
CD685768.1 | - | 2 - 3 | 248 | 148 |