Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DGAT2
Entrez Gene ID
84649
Gene Name
diacylglycerol O-acyltransferase 2
Gene Aliases
ARAT, GS1999FULL, HMFN1045
Chromosome Location
Chr.11: 75768733 - 75801536 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001253891.1 NP_001240820.1 4 - 5 770 87
NM_032564.4 NP_115953.2 5 - 6 899 87
XM_011545304.2 XP_011543606.1 5 - 6 603 87
GenBank mRNA AB048286.2 - 5 - 6 870 87
AB073384.1 - 4 - 5 447 87
AF384161.2 - 5 - 6 647 87
AK074203.1 - 1 - 2 164 87
AL834287.1 - 5 - 6 778 87
AY358532.1 - 5 - 6 824 87
AY780647.1 - 4 - 5 502 87
BC015234.1 - 5 - 6 888 87
CR749377.1 - 1 - 2 4383 87

View Full Product Details