The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163735.1 | NP_001157207.1 | 16 - 17 | 2461 | 88 |
XM_011525286.1 | XP_011523588.1 | 19 - 20 | 2148 | 88 | |
XM_011525287.1 | XP_011523589.1 | 19 - 20 | 2142 | 88 | |
XM_011525290.2 | XP_011523592.1 | 17 - 18 | 2010 | 88 | |
XM_017025157.1 | XP_016880646.1 | 19 - 20 | 2122 | 88 | |
XM_017025158.1 | XP_016880647.1 | 18 - 19 | 2055 | 88 | |
XM_017025159.1 | XP_016880648.1 | 17 - 18 | 2016 | 88 | |
XM_017025160.1 | XP_016880649.1 | 19 - 20 | 2148 | 88 | |
XM_017025161.1 | XP_016880650.1 | 17 - 18 | 1940 | 88 | |
GenBank mRNA | AB209035.1 | - | 16 - 17 | 2419 | 88 |
AK293903.1 | - | 16 - 17 | 3068 | 88 | |
AK304073.1 | - | 16 - 17 | 1812 | 88 | |
AL133017.1 | - | 2 - 3 | 2236 | 88 |