The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163735.1 | NP_001157207.1 | 21 - 22 | 3070 | 63 |
NM_025109.5 | NP_079385.2 | 17 - 18 | 2470 | 63 | |
XM_011525286.1 | XP_011523588.1 | 24 - 25 | 2757 | 63 | |
XM_011525287.1 | XP_011523589.1 | 24 - 25 | 2751 | 63 | |
XM_011525290.2 | XP_011523592.1 | 22 - 23 | 2619 | 63 | |
XM_017025157.1 | XP_016880646.1 | 24 - 25 | 2731 | 63 | |
XM_017025158.1 | XP_016880647.1 | 23 - 24 | 2664 | 63 | |
XM_017025159.1 | XP_016880648.1 | 22 - 23 | 2625 | 63 | |
XM_017025160.1 | XP_016880649.1 | 24 - 25 | 2757 | 63 | |
XM_017025161.1 | XP_016880650.1 | 22 - 23 | 2549 | 63 | |
GenBank mRNA | AK304073.1 | - | 21 - 22 | 2421 | 63 |
BC008900.2 | - | 17 - 18 | 1861 | 63 | |
BU543266.1 | - | 3 - 4 | 211 | 63 |