The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018048.1 | NP_001018058.1 | 4 - 5 | 406 | 129 |
NM_001018049.2 | NP_001018059.1 | 4 - 5 | 472 | 129 | |
NM_002571.3 | NP_002562.2 | 4 - 5 | 472 | 129 | |
XM_011518749.2 | XP_011517051.1 | 4 - 5 | 472 | 129 | |
XM_011518751.1 | XP_011517053.1 | 4 - 5 | 406 | 129 | |
GenBank mRNA | AK304657.1 | - | 4 - 5 | 406 | 129 |
AK311813.1 | - | 4 - 5 | 472 | 129 | |
BC069451.1 | - | 4 - 5 | 441 | 129 | |
BC069562.1 | - | 4 - 5 | 441 | 129 | |
BC112304.1 | - | 4 - 5 | 426 | 129 | |
BC113728.1 | - | 4 - 5 | 426 | 129 | |
DC421713.1 | - | 4 - 5 | 406 | 129 | |
J04129.1 | - | 4 - 5 | 470 | 129 | |
M61886.1 | - | 4 - 5 | 461 | 129 |