The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145293.1 | NP_001138765.1 | 5 - 6 | 983 | 77 |
NM_001145294.1 | NP_001138766.1 | 5 - 6 | 923 | 77 | |
NM_001145295.1 | NP_001138767.1 | 5 - 6 | 983 | 77 | |
NM_018297.3 | NP_060767.2 | 5 - 6 | 983 | 77 | |
XM_005265316.1 | XP_005265373.1 | 5 - 6 | 928 | 77 | |
XM_005265317.1 | XP_005265374.1 | 5 - 6 | 928 | 77 | |
XM_011533944.1 | XP_011532246.1 | 5 - 6 | 879 | 77 | |
XM_011533945.2 | XP_011532247.1 | 5 - 6 | 928 | 77 | |
XM_017006839.1 | XP_016862328.1 | 5 - 6 | 928 | 77 | |
GenBank mRNA | AF250924.2 | - | 5 - 6 | 916 | 77 |
AK296047.1 | - | 5 - 6 | 890 | 77 | |
AK310473.1 | - | 5 - 6 | 899 | 77 | |
BC000963.2 | - | 5 - 6 | 866 | 77 | |
BC007226.1 | - | 5 - 6 | 916 | 77 | |
BC017220.2 | - | 5 - 6 | 910 | 77 |