The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145293.1 | NP_001138765.1 | 8 - 9 | 1312 | 134 |
NM_001145294.1 | NP_001138766.1 | 8 - 9 | 1306 | 134 | |
NM_001145295.1 | NP_001138767.1 | 8 - 9 | 1366 | 134 | |
NM_018297.3 | NP_060767.2 | 8 - 9 | 1366 | 134 | |
XM_011533944.1 | XP_011532246.1 | 8 - 9 | 1262 | 134 | |
XM_017006839.1 | XP_016862328.1 | 8 - 9 | 1311 | 134 | |
GenBank mRNA | AF250924.2 | - | 8 - 9 | 1299 | 134 |
AK001867.1 | - | 6 - 7 | 967 | 134 | |
AK296047.1 | - | 8 - 9 | 1273 | 134 | |
AK310473.1 | - | 7 - 8 | 2161 | 134 | |
BC000963.2 | - | 8 - 9 | 1195 | 134 | |
BC007226.1 | - | 8 - 9 | 1299 | 134 | |
BC017220.2 | - | 8 - 9 | 1293 | 134 |