Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RTTN
Entrez Gene ID
25914
Gene Name
rotatin
Gene Aliases
MSSP
Chromosome Location
Chr.18: 70003804 - 70205726 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318520.1 NP_001305449.1 5 - 6 639 80
NM_173630.3 NP_775901.3 5 - 6 639 80
XM_006722434.3 XP_006722497.1 5 - 6 639 80
XM_006722435.3 XP_006722498.1 5 - 6 639 80
XM_011525902.2 XP_011524204.1 5 - 6 639 80
XM_011525903.2 XP_011524205.1 5 - 6 639 80
XM_011525904.2 XP_011524206.1 5 - 6 639 80
XM_011525905.2 XP_011524207.1 5 - 6 639 80
XM_011525907.2 XP_011524209.1 5 - 6 639 80
XM_011525908.2 XP_011524210.1 5 - 6 639 80
XM_017025693.1 XP_016881182.1 5 - 6 639 80
XM_017025694.1 XP_016881183.1 2 - 3 194 80
XM_017025696.1 XP_016881185.1 5 - 6 574 80
GenBank mRNA AK096404.1 - 5 - 6 642 80
BC143723.1 - 5 - 6 613 80

View Full Product Details