The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135729.1 | NP_001129201.1 | 5 - 6 | 548 | 78 |
NM_001135732.1 | NP_001129204.1 | 5 - 6 | 621 | 78 | |
NM_005488.2 | NP_005479.1 | 5 - 6 | 621 | 78 | |
NR_024194.1 | 5 - 6 | 650 | 78 | ||
NR_024195.1 | 4 - 5 | 571 | 78 | ||
XM_011529818.2 | XP_011528120.1 | 6 - 7 | 635 | 78 | |
XM_011529820.2 | XP_011528122.1 | 4 - 5 | 486 | 78 | |
XM_017028529.1 | XP_016884018.1 | 6 - 7 | 635 | 78 | |
XM_017028530.1 | XP_016884019.1 | 4 - 5 | 486 | 78 | |
GenBank mRNA | AJ006973.1 | - | 5 - 6 | 557 | 78 |
AK026576.1 | - | 5 - 6 | 574 | 78 | |
AK097926.1 | - | 6 - 7 | 637 | 78 | |
AK293694.1 | - | 5 - 6 | 548 | 78 | |
AK295918.1 | - | 4 - 5 | 481 | 78 | |
AK296670.1 | - | 5 - 6 | 558 | 78 | |
AK314522.1 | - | 5 - 6 | 530 | 78 | |
BC040006.1 | - | 5 - 6 | 514 | 78 | |
BC046151.1 | - | 5 - 6 | 499 | 78 | |
CR456595.1 | - | 5 - 6 | 517 | 78 |