The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278585.1 | NP_001265514.1 | 17 - 18 | 2384 | 80 |
NM_006587.3 | NP_006578.2 | 19 - 20 | 2696 | 80 | |
GenBank mRNA | AF133845.1 | - | 19 - 20 | 2632 | 80 |
AK302816.1 | - | 18 - 19 | 2371 | 80 | |
AK304038.1 | - | 18 - 19 | 2461 | 80 | |
BC110451.2 | - | 19 - 20 | 2632 | 80 |