The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278585.1 | NP_001265514.1 | 8 - 9 | 1197 | 91 |
NM_001278586.1 | NP_001265515.1 | 9 - 10 | 1398 | 91 | |
NM_006587.3 | NP_006578.2 | 10 - 11 | 1509 | 91 | |
GenBank mRNA | AF133845.1 | - | 10 - 11 | 1445 | 91 |
AK297675.1 | - | 9 - 10 | 1359 | 91 | |
AK302816.1 | - | 9 - 10 | 1184 | 91 | |
AK304038.1 | - | 9 - 10 | 1274 | 91 | |
AK304466.1 | - | 9 - 10 | 1398 | 91 | |
BC110451.2 | - | 10 - 11 | 1445 | 91 |