The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142633.2 | NP_001136105.1 | 10 - 11 | 1749 | 53 |
NM_001251851.1 | NP_001238780.1 | 10 - 11 | 1811 | 53 | |
NM_001251852.1 | NP_001238781.1 | 9 - 10 | 1633 | 53 | |
NM_001251853.1 | NP_001238782.1 | 9 - 10 | 1595 | 53 | |
NM_001251855.1 | NP_001238784.1 | 9 - 10 | 1574 | 53 | |
NM_014308.3 | NP_055123.2 | 10 - 11 | 1686 | 53 | |
XM_005256580.3 | XP_005256637.1 | 10 - 11 | 1744 | 53 | |
GenBank mRNA | AB028925.1 | - | 9 - 10 | 1619 | 53 |
AF128881.1 | - | 10 - 11 | 1686 | 53 | |
AK314186.1 | - | 10 - 11 | 1744 | 53 | |
BC028212.1 | - | 9 - 10 | 1565 | 53 |