The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142633.2 | NP_001136105.1 | 18 - 19 | 2618 | 68 |
NM_001251851.1 | NP_001238780.1 | 18 - 19 | 2680 | 68 | |
NM_001251852.1 | NP_001238781.1 | 17 - 18 | 2502 | 68 | |
NM_001251853.1 | NP_001238782.1 | 17 - 18 | 2464 | 68 | |
NM_001251855.1 | NP_001238784.1 | 17 - 18 | 2443 | 68 | |
NM_014308.3 | NP_055123.2 | 18 - 19 | 2555 | 68 | |
XM_005256580.3 | XP_005256637.1 | 18 - 19 | 2610 | 68 | |
GenBank mRNA | AB028925.1 | - | 17 - 18 | 2488 | 68 |
AF128881.1 | - | 18 - 19 | 2555 | 68 | |
AK314186.1 | - | 17 - 18 | 2685 | 68 | |
BC028212.1 | - | 17 - 18 | 2434 | 68 |