The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142633.2 | NP_001136105.1 | 4 - 5 | 407 | 78 |
NM_001251851.1 | NP_001238780.1 | 4 - 5 | 407 | 78 | |
NM_001251852.1 | NP_001238781.1 | 3 - 4 | 291 | 78 | |
NM_001251853.1 | NP_001238782.1 | 4 - 5 | 407 | 78 | |
NM_001251855.1 | NP_001238784.1 | 4 - 5 | 407 | 78 | |
NM_014308.3 | NP_055123.2 | 4 - 5 | 344 | 78 | |
XM_005256580.3 | XP_005256637.1 | 4 - 5 | 402 | 78 | |
GenBank mRNA | AB028925.1 | - | 3 - 4 | 277 | 78 |
AF128881.1 | - | 4 - 5 | 344 | 78 | |
AK309074.1 | - | 3 - 4 | 291 | 78 | |
AK314186.1 | - | 4 - 5 | 402 | 78 | |
AY725851.1 | - | 4 - 5 | 310 | 78 | |
AY725853.1 | - | 4 - 5 | 310 | 78 | |
AY725854.1 | - | 4 - 5 | 310 | 78 | |
BC028212.1 | - | 4 - 5 | 398 | 78 |