Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCAT1
Entrez Gene ID
586
Gene Name
branched chain amino acid transaminase 1
Gene Aliases
BCATC, BCT1, ECA39, MECA39, PNAS121, PP18
Chromosome Location
Chr.12: 24810024 - 24949459 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178091.1 NP_001171562.1 6 - 7 1234 122
NM_001178092.1 NP_001171563.1 5 - 6 1162 122
NM_001178093.1 NP_001171564.1 7 - 8 941 122
NM_001178094.1 NP_001171565.1 7 - 8 1238 122
NM_005504.6 NP_005495.2 7 - 8 1345 122
XM_017019768.1 XP_016875257.1 6 - 7 953 122
GenBank mRNA AK056255.1 - 7 - 8 1345 122
AK128527.1 - 5 - 6 789 122
AK294879.1 - 7 - 8 924 122
AK299088.1 - 6 - 7 818 122
BX647122.1 - 7 - 8 947 122
CR749308.1 - 7 - 8 1147 122

View Full Product Details