Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STRA8
Entrez Gene ID
346673
Gene Name
stimulated by retinoic acid 8
Gene Aliases
-
Chromosome Location
Chr.7: 135231979 - 135258661 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182489.1 NP_872295.1 7 - 8 811 69
XM_011516137.2 XP_011514439.1 7 - 8 1664 69
XM_011516138.2 XP_011514440.1 7 - 8 1049 69
XM_011516139.2 XP_011514441.1 8 - 9 1293 69
GenBank mRNA AF513502.1 - 7 - 8 811 69

View Full Product Details