Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ARMC8
- Entrez Gene ID
-
25852
- Gene Name
- armadillo repeat containing 8
- Gene Aliases
-
GID5, HSPC056, S863-2, VID28
- Chromosome Location
-
Chr.3: 138187248 - 138298389 on Build GRCh38
View Full Product Details