The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000655.4 | NP_000646.2 | 5 - 6 | 963 | 75 |
NR_029467.1 | 3 - 4 | 727 | 75 | ||
GenBank mRNA | AB583235.1 | - | 4 - 5 | 759 | 75 |
AJ246000.1 | - | 5 - 6 | 834 | 75 | |
AK225713.1 | - | 5 - 6 | 874 | 75 | |
AK298705.1 | - | 3 - 4 | 722 | 75 | |
AK312673.1 | - | 5 - 6 | 874 | 75 | |
AY367061.1 | - | 3 - 4 | 327 | 75 | |
BC020758.1 | - | 5 - 6 | 860 | 75 | |
M25280.1 | - | 5 - 6 | 874 | 75 | |
X16070.1 | - | 5 - 6 | 887 | 75 | |
X17519.1 | - | 5 - 6 | 847 | 75 |