The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018429.2 | NP_060899.2 | 14 - 15 | 2310 | 70 |
XM_011543511.2 | XP_011541813.1 | 14 - 15 | 2288 | 70 | |
XM_011543512.2 | XP_011541814.1 | 14 - 15 | 2289 | 70 | |
XM_017009630.1 | XP_016865119.1 | 14 - 15 | 2289 | 70 | |
XM_017009631.1 | XP_016865120.1 | 14 - 15 | 2289 | 70 | |
XM_017009632.1 | XP_016865121.1 | 14 - 15 | 2288 | 70 | |
XM_017009633.1 | XP_016865122.1 | 14 - 15 | 2288 | 70 | |
XM_017009634.1 | XP_016865123.1 | 14 - 15 | 2288 | 70 | |
GenBank mRNA | AB033067.1 | - | - | 1230 | 70 |
AB051476.3 | - | 14 - 15 | 2057 | 70 | |
AF176695.1 | - | 14 - 15 | 2266 | 70 | |
AF298151.1 | - | 14 - 15 | 2136 | 70 | |
AF298152.1 | - | 14 - 15 | 2136 | 70 | |
AJ238520.2 | - | 14 - 15 | 2251 | 70 | |
BC146792.1 | - | 14 - 15 | 2057 | 70 | |
BC161615.1 | - | 14 - 15 | 2039 | 70 |