Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GTPBP10
Entrez Gene ID
85865
Gene Name
GTP binding protein 10
Gene Aliases
ObgH2, UG0751c10
Chromosome Location
Chr.7: 90346665 - 90391455 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042717.2 NP_001036182.1 2 - 3 300 88
GenBank mRNA AF351613.1 - 2 - 3 301 88
AK294325.1 - 2 - 3 250 88
BC144162.1 - 2 - 3 247 88

View Full Product Details