The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256725.1 | NP_001243654.1 | 5 - 6 | 824 | 137 |
NM_001256726.1 | NP_001243655.1 | 5 - 6 | 824 | 137 | |
NM_001256727.1 | NP_001243656.1 | 5 - 6 | 709 | 137 | |
NM_001256728.1 | NP_001243657.1 | 4 - 5 | 634 | 137 | |
NM_006590.3 | NP_006581.2 | 5 - 6 | 824 | 137 | |
NR_046347.1 | 5 - 6 | 712 | 137 | ||
XM_006711922.1 | XP_006711985.1 | 5 - 6 | 692 | 137 | |
XM_006711923.2 | XP_006711986.1 | 5 - 6 | 771 | 137 | |
XM_011532487.1 | XP_011530789.1 | 5 - 6 | 777 | 137 | |
XM_011532488.1 | XP_011530790.1 | 5 - 6 | 710 | 137 | |
XM_017003182.1 | XP_016858671.1 | 5 - 6 | 781 | 137 | |
GenBank mRNA | AF130096.1 | - | 4 - 5 | 413 | 137 |
AF132955.1 | - | 5 - 6 | 729 | 137 | |
AF353989.1 | - | 5 - 6 | 731 | 137 | |
AK001047.1 | - | 5 - 6 | 722 | 137 | |
AK001772.1 | - | 1 - 2 | 117 | 137 | |
AK056334.1 | - | 5 - 6 | 693 | 137 | |
AK129872.1 | - | 6 - 7 | 705 | 137 | |
AK222852.1 | - | 5 - 6 | 727 | 137 | |
AK289451.1 | - | 5 - 6 | 778 | 137 | |
AK295257.1 | - | 4 - 5 | 632 | 137 | |
AK302236.1 | - | 5 - 6 | 686 | 137 | |
AK309485.1 | - | 6 - 7 | 627 | 137 | |
AL117416.1 | - | 3 - 4 | 391 | 137 | |
AL519827.3 | - | 5 - 6 | 774 | 137 | |
BC001384.2 | - | 5 - 6 | 713 | 137 | |
BC067273.1 | - | 5 - 6 | 752 | 137 | |
BX462623.2 | - | 5 - 6 | 757 | 137 |