The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256725.1 | NP_001243654.1 | 8 - 9 | 1198 | 88 |
NM_001256726.1 | NP_001243655.1 | 8 - 9 | 1198 | 88 | |
NM_001256727.1 | NP_001243656.1 | 8 - 9 | 1083 | 88 | |
NM_001256728.1 | NP_001243657.1 | 7 - 8 | 1008 | 88 | |
NM_006590.3 | NP_006581.2 | 8 - 9 | 1198 | 88 | |
NR_046347.1 | 8 - 9 | 1086 | 88 | ||
XM_006711922.1 | XP_006711985.1 | 8 - 9 | 1066 | 88 | |
XM_006711923.2 | XP_006711986.1 | 8 - 9 | 1145 | 88 | |
XM_011532487.1 | XP_011530789.1 | 8 - 9 | 1151 | 88 | |
XM_011532488.1 | XP_011530790.1 | 8 - 9 | 1084 | 88 | |
XM_017003182.1 | XP_016858671.1 | 8 - 9 | 1155 | 88 | |
GenBank mRNA | AF130096.1 | - | 8 - 9 | 787 | 88 |
AF132955.1 | - | 8 - 9 | 1103 | 88 | |
AF161450.1 | - | 3 - 4 | 184 | 88 | |
AF353989.1 | - | 8 - 9 | 1105 | 88 | |
AK001047.1 | - | 8 - 9 | 1096 | 88 | |
AK001772.1 | - | 4 - 5 | 491 | 88 | |
AK056334.1 | - | 8 - 9 | 1067 | 88 | |
AK129872.1 | - | 9 - 10 | 1079 | 88 | |
AK222852.1 | - | 8 - 9 | 1101 | 88 | |
AK289451.1 | - | 8 - 9 | 1152 | 88 | |
AK295257.1 | - | 7 - 8 | 1006 | 88 | |
AK302236.1 | - | 8 - 9 | 1060 | 88 | |
AK309485.1 | - | 10 - 11 | 1001 | 88 | |
AL117416.1 | - | 6 - 7 | 765 | 88 | |
BC001384.2 | - | 8 - 9 | 1087 | 88 | |
BC067273.1 | - | 8 - 9 | 1126 | 88 |