Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR75
Entrez Gene ID
84128
Gene Name
WD repeat domain 75
Gene Aliases
NET16, UTP17
Chromosome Location
Chr.2: 189441433 - 189475562 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303096.1 NP_001290025.1 19 - 20 2147 98
NM_032168.2 NP_115544.1 18 - 19 2113 98
GenBank mRNA AK022581.1 - 18 - 19 2060 98
AK022607.1 - 18 - 19 2072 98
AK023276.1 - 15 - 16 1752 98
AK091546.1 - 19 - 20 2074 98
AK290445.1 - 18 - 19 2064 98
AL122097.1 - 14 - 15 1680 98
BC006816.1 - 3 - 4 307 98
BC018689.1 - 1 - 2 272 98
BC040567.1 - 18 - 19 2113 98
BU619995.1 - 3 - 4 97
CR749440.1 - 18 - 19 3722 98

View Full Product Details