Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR75
Entrez Gene ID
84128
Gene Name
WD repeat domain 75
Gene Aliases
NET16, UTP17
Chromosome Location
Chr.2: 189441433 - 189475562 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303096.1 NP_001290025.1 5 - 6 468 106
NM_032168.2 NP_115544.1 4 - 5 434 106
GenBank mRNA AK022607.1 - 4 - 5 393 106
AK091546.1 - 5 - 6 395 106
AK290445.1 - 4 - 5 385 106
BC040567.1 - 4 - 5 434 106
BI460450.1 - 4 - 5 439 106
CR749440.1 - 4 - 5 2043 106

View Full Product Details