Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR75
Entrez Gene ID
84128
Gene Name
WD repeat domain 75
Gene Aliases
NET16, UTP17
Chromosome Location
Chr.2: 189441433 - 189475562 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303096.1 NP_001290025.1 7 - 8 665 135
NM_032168.2 NP_115544.1 6 - 7 631 135
GenBank mRNA AK022581.1 - 6 - 7 578 135
AK022607.1 - 6 - 7 590 135
AK023276.1 - 3 - 4 270 135
AK091546.1 - 7 - 8 592 135
AK290445.1 - 6 - 7 582 135
AL122097.1 - 2 - 3 198 135
BC040567.1 - 6 - 7 631 135
CR749440.1 - 6 - 7 2240 135

View Full Product Details