Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INPP4A
Entrez Gene ID
3631
Gene Name
inositol polyphosphate-4-phosphatase type I A
Gene Aliases
INPP4, TVAS1
Chromosome Location
Chr.2: 98444592 - 98594068 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134224.1 NP_001127696.1 16 - 17 1971 86
NM_001134225.1 NP_001127697.1 15 - 16 1956 86
NM_001566.2 NP_001557.1 16 - 17 1971 86
NM_004027.2 NP_004018.1 16 - 17 1971 86
XM_006712499.3 XP_006712562.1 16 - 17 1969 86
XM_006712500.3 XP_006712563.1 16 - 17 1966 86
XM_006712501.3 XP_006712564.1 15 - 16 1951 86
XM_006712502.3 XP_006712565.1 16 - 17 1969 86
XM_006712503.3 XP_006712566.1 16 - 17 1969 86
XM_006712504.3 XP_006712567.1 16 - 17 1969 86
XM_006712505.3 XP_006712568.1 16 - 17 1969 86
XM_006712506.3 XP_006712569.1 16 - 17 1969 86
XM_006712507.3 XP_006712570.1 16 - 17 1969 86
XM_006712508.3 XP_006712571.1 16 - 17 1969 86
XM_011511122.2 XP_011509424.1 15 - 16 1919 86
XM_011511123.2 XP_011509425.1 16 - 17 1969 86
XM_011511124.2 XP_011509426.1 15 - 16 1951 86
XM_011511125.2 XP_011509427.1 13 - 14 1851 86
XM_011511127.2 XP_011509429.1 4 - 5 654 86
XM_017003993.1 XP_016859482.1 16 - 17 1810 86
XM_017003994.1 XP_016859483.1 15 - 16 1951 86
XM_017003995.1 XP_016859484.1 16 - 17 1966 86
XM_017003996.1 XP_016859485.1 16 - 17 1969 86
XM_017003997.1 XP_016859486.1 16 - 17 2028 86
XM_017003998.1 XP_016859487.1 16 - 17 2025 86
XM_017003999.1 XP_016859488.1 15 - 16 2009 86
XM_017004000.1 XP_016859489.1 16 - 17 1969 86
XM_017004001.1 XP_016859490.1 15 - 16 1951 86
XM_017004002.1 XP_016859491.1 15 - 16 1869 86
XM_017004003.1 XP_016859492.1 16 - 17 1969 86
GenBank mRNA AF368319.1 - 14 - 15 1578 86
BC028361.2 - 15 - 16 1956 86
U26398.1 - 16 - 17 1872 86
U96919.1 - 16 - 17 1872 86

View Full Product Details