Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INPP4A
Entrez Gene ID
3631
Gene Name
inositol polyphosphate-4-phosphatase type I A
Gene Aliases
INPP4, TVAS1
Chromosome Location
Chr.2: 98444592 - 98594068 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134224.1 NP_001127696.1 19 - 20 2441 59
NM_001134225.1 NP_001127697.1 18 - 19 2426 59
NM_001566.2 NP_001557.1 19 - 20 2324 59
NM_004027.2 NP_004018.1 19 - 20 2324 59
XM_006712499.3 XP_006712562.1 19 - 20 2439 59
XM_006712500.3 XP_006712563.1 19 - 20 2436 59
XM_006712501.3 XP_006712564.1 18 - 19 2421 59
XM_006712502.3 XP_006712565.1 18 - 19 2406 59
XM_006712503.3 XP_006712566.1 19 - 20 2439 59
XM_006712504.3 XP_006712567.1 19 - 20 2322 59
XM_006712505.3 XP_006712568.1 19 - 20 2439 59
XM_006712506.3 XP_006712569.1 18 - 19 2289 59
XM_006712507.3 XP_006712570.1 19 - 20 2322 59
XM_006712508.3 XP_006712571.1 19 - 20 2439 59
XM_011511122.2 XP_011509424.1 18 - 19 2389 59
XM_011511123.2 XP_011509425.1 19 - 20 2439 59
XM_011511124.2 XP_011509426.1 18 - 19 2304 59
XM_011511125.2 XP_011509427.1 16 - 17 2321 59
XM_011511127.2 XP_011509429.1 7 - 8 1124 59
XM_017003993.1 XP_016859482.1 19 - 20 2163 59
XM_017003994.1 XP_016859483.1 18 - 19 2304 59
XM_017003995.1 XP_016859484.1 18 - 19 2286 59
XM_017003996.1 XP_016859485.1 19 - 20 2439 59
XM_017003997.1 XP_016859486.1 19 - 20 2381 59
XM_017003998.1 XP_016859487.1 19 - 20 2378 59
XM_017003999.1 XP_016859488.1 18 - 19 2362 59
XM_017004000.1 XP_016859489.1 18 - 19 2289 59
XM_017004001.1 XP_016859490.1 17 - 18 2271 59
XM_017004002.1 XP_016859491.1 17 - 18 2189 59
XM_017004003.1 XP_016859492.1 19 - 20 2322 59
GenBank mRNA AF368319.1 - 17 - 18 2048 59
BC028361.2 - 18 - 19 2426 59
U26398.1 - 19 - 20 2225 59
U96919.1 - 19 - 20 2225 59

View Full Product Details