Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INPP4A
Entrez Gene ID
3631
Gene Name
inositol polyphosphate-4-phosphatase type I A
Gene Aliases
INPP4, TVAS1
Chromosome Location
Chr.2: 98444592 - 98594068 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134224.1 NP_001127696.1 20 - 21 2557 70
NM_001134225.1 NP_001127697.1 19 - 20 2542 70
NM_001566.2 NP_001557.1 20 - 21 2440 70
NM_004027.2 NP_004018.1 20 - 21 2440 70
XM_006712499.3 XP_006712562.1 20 - 21 2555 70
XM_006712500.3 XP_006712563.1 20 - 21 2552 70
XM_006712501.3 XP_006712564.1 19 - 20 2537 70
XM_006712502.3 XP_006712565.1 19 - 20 2522 70
XM_006712503.3 XP_006712566.1 20 - 21 2555 70
XM_006712504.3 XP_006712567.1 20 - 21 2438 70
XM_006712505.3 XP_006712568.1 20 - 21 2555 70
XM_006712506.3 XP_006712569.1 19 - 20 2405 70
XM_006712507.3 XP_006712570.1 20 - 21 2438 70
XM_006712508.3 XP_006712571.1 20 - 21 2555 70
XM_011511122.2 XP_011509424.1 19 - 20 2505 70
XM_011511123.2 XP_011509425.1 20 - 21 2555 70
XM_011511124.2 XP_011509426.1 19 - 20 2420 70
XM_011511125.2 XP_011509427.1 17 - 18 2437 70
XM_011511127.2 XP_011509429.1 8 - 9 1240 70
XM_017003993.1 XP_016859482.1 20 - 21 2279 70
XM_017003994.1 XP_016859483.1 19 - 20 2420 70
XM_017003995.1 XP_016859484.1 19 - 20 2402 70
XM_017003996.1 XP_016859485.1 20 - 21 2555 70
XM_017003997.1 XP_016859486.1 20 - 21 2497 70
XM_017003998.1 XP_016859487.1 20 - 21 2494 70
XM_017003999.1 XP_016859488.1 19 - 20 2478 70
XM_017004000.1 XP_016859489.1 19 - 20 2405 70
XM_017004001.1 XP_016859490.1 18 - 19 2387 70
XM_017004002.1 XP_016859491.1 18 - 19 2305 70
XM_017004003.1 XP_016859492.1 20 - 21 2438 70
GenBank mRNA AF368319.1 - 18 - 19 2164 70
BC028361.2 - 19 - 20 2542 70
U26398.1 - 20 - 21 2341 70
U96919.1 - 20 - 21 2341 70

View Full Product Details