Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- INPP4A
- Entrez Gene ID
-
3631
- Gene Name
- inositol polyphosphate-4-phosphatase type I A
- Gene Aliases
-
INPP4, TVAS1
- Chromosome Location
-
Chr.2: 98444592 - 98594068 on Build GRCh38
View Full Product Details