Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCNF
Entrez Gene ID
899
Gene Name
cyclin F
Gene Aliases
FBX1, FBXO1
Chromosome Location
Chr.16: 2425438 - 2458858 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001323538.1 NP_001310467.1 10 - 11 1209 61
NM_001761.2 NP_001752.2 11 - 12 1313 61
XM_017023822.1 XP_016879311.1 11 - 12 1473 61
GenBank mRNA AB208829.1 - 10 - 11 1120 61
AK313371.1 - 11 - 12 1266 61
BC012349.1 - 11 - 12 1237 61
U17105.1 - 11 - 12 1238 61
Z36714.1 - 11 - 12 1268 61

View Full Product Details