Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCNF
Entrez Gene ID
899
Gene Name
cyclin F
Gene Aliases
FBX1, FBXO1
Chromosome Location
Chr.16: 2425438 - 2458858 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001323538.1 NP_001310467.1 5 - 6 579 79
NM_001761.2 NP_001752.2 6 - 7 683 79
XM_017023822.1 XP_016879311.1 6 - 7 843 79
XM_017023823.1 XP_016879312.1 6 - 7 843 79
GenBank mRNA AB208829.1 - 5 - 6 490 79
AK313371.1 - 6 - 7 636 79
BC012349.1 - 6 - 7 607 79
U17105.1 - 6 - 7 608 79
Z36714.1 - 6 - 7 638 79

View Full Product Details