The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127.3 | NP_001118.3 | 13 - 14 | 1984 | 69 |
NM_001166019.1 | NP_001159491.1 | 13 - 14 | 1984 | 69 | |
NM_145730.2 | NP_663782.2 | 13 - 14 | 1984 | 69 | |
GenBank mRNA | AK294406.1 | - | 4 - 5 | 548 | 69 |
AK296567.1 | - | 2 - 3 | 328 | 69 | |
AK299613.1 | - | 4 - 5 | 549 | 69 | |
BC046242.1 | - | 13 - 14 | 1984 | 69 | |
CT841508.1 | - | 12 - 13 | 1827 | 69 | |
L13939.1 | - | 12 - 13 | 1843 | 69 |