Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AP1B1
Entrez Gene ID
162
Gene Name
adaptor related protein complex 1 beta 1 subunit
Gene Aliases
ADTB1, AP105A, BAM22, CLAPB2
Chromosome Location
Chr.22: 29327680 - 29388583 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127.3 NP_001118.3 20 - 21 2796 86
NM_001166019.1 NP_001159491.1 19 - 20 2715 86
NM_145730.2 NP_663782.2 19 - 20 2775 86
GenBank mRNA AK294406.1 - 11 - 12 86
AK296567.1 - 8 - 9 1119 86
AK299613.1 - 11 - 12 1361 86
AU139022.1 - 4 - 5 377 86
BC046242.1 - 19 - 20 2715 86
BX537680.1 - 3 - 4 705 86
CT841508.1 - 18 - 19 2618 86
L13939.1 - 19 - 20 2655 86

View Full Product Details