The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004274.4 | NP_004265.3 | 12 - 13 | 3757 | 75 |
XM_011537378.2 | XP_011535680.1 | 7 - 8 | 1221 | 75 | |
XM_011537379.2 | XP_011535681.1 | 6 - 7 | 1158 | 75 | |
XM_011537383.2 | XP_011535685.1 | 4 - 5 | 609 | 75 | |
XM_017021808.1 | XP_016877297.1 | 11 - 12 | 3624 | 75 | |
XM_017021809.1 | XP_016877298.1 | 11 - 12 | 4997 | 75 | |
XM_017021810.1 | XP_016877299.1 | 12 - 13 | 5187 | 75 | |
XM_017021811.1 | XP_016877300.1 | 6 - 7 | 6898 | 75 | |
GenBank mRNA | AB002309.2 | - | 12 - 13 | 3697 | 75 |
BC137232.1 | - | 12 - 13 | 3640 | 75 | |
BC137233.1 | - | 12 - 13 | 3640 | 75 | |
BC150288.1 | - | 12 - 13 | 3697 | 75 | |
U17195.2 | - | 12 - 13 | 3713 | 75 |