The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004274.4 | NP_004265.3 | 8 - 9 | 3052 | 113 |
XM_011537378.2 | XP_011535680.1 | 3 - 4 | 516 | 113 | |
XM_011537379.2 | XP_011535681.1 | 2 - 3 | 453 | 113 | |
XM_017021808.1 | XP_016877297.1 | 7 - 8 | 2919 | 113 | |
XM_017021809.1 | XP_016877298.1 | 7 - 8 | 4292 | 113 | |
XM_017021810.1 | XP_016877299.1 | 8 - 9 | 4482 | 113 | |
XM_017021811.1 | XP_016877300.1 | 2 - 3 | 6193 | 113 | |
GenBank mRNA | AB002309.2 | - | 8 - 9 | 2992 | 113 |
BC137232.1 | - | 8 - 9 | 2935 | 113 | |
BC137233.1 | - | 8 - 9 | 2935 | 113 | |
BC150185.1 | - | 8 - 9 | 2981 | 113 | |
BC150288.1 | - | 8 - 9 | 2992 | 113 | |
BC154413.1 | - | 8 - 9 | 2981 | 113 |