Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAP7D1
Entrez Gene ID
55700
Gene Name
MAP7 domain containing 1
Gene Aliases
PARCC1, RPRC1
Chromosome Location
Chr.1: 36155965 - 36180849 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286365.1 NP_001273294.1 8 - 9 2052 111
NM_001286366.1 NP_001273295.1 10 - 11 2067 111
NM_018067.4 NP_060537.3 9 - 10 2163 111
XM_005271027.1 XP_005271084.1 8 - 9 1867 111
XM_006710759.1 XP_006710822.1 9 - 10 1978 111
XM_006710761.1 XP_006710824.1 9 - 10 1771 111
GenBank mRNA AB033013.2 - 8 - 9 2052 111
AF218002.1 - 9 - 10 1389 111
AK001212.1 - 1 - 2 246 111
AK096341.1 - 10 - 11 1834 111
AK309966.1 - 7 - 8 1058 111
AL136547.1 - 1 - 2 77 111
AY007099.1 - 1 - 2 292 111
BC003083.2 - 1 - 2 313 111
BC057293.1 - 8 - 9 1854 111
BC067256.1 - 6 - 7 1225 111
BC082266.1 - 3 - 4 588 111
BC106053.1 - 9 - 10 1926 111

View Full Product Details