The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286365.1 | NP_001273294.1 | 7 - 8 | 1727 | 64 |
NM_001286366.1 | NP_001273295.1 | 9 - 10 | 1742 | 64 | |
NM_018067.4 | NP_060537.3 | 8 - 9 | 1838 | 64 | |
XM_005271027.1 | XP_005271084.1 | 7 - 8 | 1542 | 64 | |
XM_006710759.1 | XP_006710822.1 | 8 - 9 | 1653 | 64 | |
XM_006710761.1 | XP_006710824.1 | 8 - 9 | 1446 | 64 | |
GenBank mRNA | AB033013.2 | - | 7 - 8 | 1727 | 64 |
AF218002.1 | - | 8 - 9 | 1064 | 64 | |
AK096341.1 | - | 9 - 10 | 1509 | 64 | |
AK309966.1 | - | 6 - 7 | 733 | 64 | |
BC057293.1 | - | 7 - 8 | 1529 | 64 | |
BC067256.1 | - | 5 - 6 | 900 | 64 | |
BC082266.1 | - | 2 - 3 | 263 | 64 | |
BC106053.1 | - | 8 - 9 | 1601 | 64 |