Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAMTS18
Entrez Gene ID
170692
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 18
Gene Aliases
ADAMTS21, KNO2, MMCAT
Chromosome Location
Chr.16: 77282128 - 77435114 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001326358.1 NP_001313287.1 11 - 12 2136 73
NM_199355.3 NP_955387.1 11 - 12 2132 73
XM_011522923.1 XP_011521225.1 9 - 10 1543 73
XM_011522924.1 XP_011521226.1 9 - 10 1539 73
XM_017022988.1 XP_016878477.1 8 - 9 1379 73
XM_017022989.1 XP_016878478.1 8 - 9 1176 73
GenBank mRNA AJ311903.1 - 11 - 12 1792 73
AK131403.1 - 10 - 11 1639 73
BC063283.1 - 11 - 12 2132 73

View Full Product Details