Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAMTS18
Entrez Gene ID
170692
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 18
Gene Aliases
ADAMTS21, KNO2, MMCAT
Chromosome Location
Chr.16: 77282128 - 77435114 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001326358.1 NP_001313287.1 12 - 13 2283 69
NM_199355.3 NP_955387.1 12 - 13 2279 69
XM_011522923.1 XP_011521225.1 10 - 11 1690 69
XM_011522924.1 XP_011521226.1 10 - 11 1686 69
XM_017022988.1 XP_016878477.1 9 - 10 1526 69
XM_017022989.1 XP_016878478.1 9 - 10 1323 69
GenBank mRNA AJ311903.1 - 12 - 13 1939 69
AK131403.1 - 11 - 12 1786 69
BC063283.1 - 12 - 13 2279 69

View Full Product Details