Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAMTS18
Entrez Gene ID
170692
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 18
Gene Aliases
ADAMTS21, KNO2, MMCAT
Chromosome Location
Chr.16: 77282128 - 77435114 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001326358.1 NP_001313287.1 18 - 19 3228 69
NM_199355.3 NP_955387.1 18 - 19 3224 69
XM_011522923.1 XP_011521225.1 16 - 17 2635 69
XM_011522924.1 XP_011521226.1 16 - 17 2631 69
XM_017022988.1 XP_016878477.1 15 - 16 2471 69
XM_017022989.1 XP_016878478.1 15 - 16 2268 69
GenBank mRNA AJ311903.1 - 18 - 19 2884 69
AK122677.1 - 3 - 4 525 69
AY172951.1 - 3 - 4 303 69
BC063283.1 - 18 - 19 3224 69

View Full Product Details