Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADAMTS18
Entrez Gene ID
170692
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 18
Gene Aliases
ADAMTS21, KNO2, MMCAT
Chromosome Location
Chr.16: 77282128 - 77435114 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001326358.1 NP_001313287.1 5 - 6 1396 103
NM_199355.3 NP_955387.1 5 - 6 1392 103
XM_011522923.1 XP_011521225.1 3 - 4 803 103
XM_011522924.1 XP_011521226.1 3 - 4 799 103
XM_017022988.1 XP_016878477.1 3 - 4 799 103
XM_017022989.1 XP_016878478.1 3 - 4 596 103
GenBank mRNA AK293831.1 - 5 - 6 1316 103
BC063283.1 - 5 - 6 1392 103

View Full Product Details