The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003825.3 | NP_003816.2 | 5 - 6 | 682 | 68 |
XM_017022693.1 | XP_016878182.1 | 5 - 6 | 388 | 68 | |
XM_017022694.1 | XP_016878183.1 | 6 - 7 | 450 | 68 | |
GenBank mRNA | AJ011915.1 | - | 5 - 6 | 304 | 68 |
AK290152.1 | - | 5 - 6 | 728 | 68 | |
AK313161.1 | - | 5 - 6 | 326 | 68 | |
BC000148.2 | - | 5 - 6 | 314 | 68 | |
BC003686.1 | - | 5 - 6 | 333 | 68 | |
BC022890.1 | - | 5 - 6 | 338 | 68 | |
BI918294.1 | - | 5 - 6 | 350 | 68 | |
BT006916.1 | - | 4 - 5 | 260 | 68 | |
CR457212.1 | - | 4 - 5 | 260 | 68 | |
U55936.1 | - | 5 - 6 | 352 | 68 | |
Y09567.1 | - | 4 - 5 | 260 | 68 |