Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNAP23
Entrez Gene ID
8773
Gene Name
synaptosome associated protein 23
Gene Aliases
HsT17016, SNAP-23, SNAP23A, SNAP23B
Chromosome Location
Chr.15: 42491244 - 42533061 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003825.3 NP_003816.2 6 - 7 849 80
XM_006720725.2 XP_006720788.1 7 - 8 928 80
XM_017022692.1 XP_016878181.1 7 - 8 928 80
XM_017022693.1 XP_016878182.1 6 - 7 555 80
XM_017022694.1 XP_016878183.1 7 - 8 617 80
GenBank mRNA AJ011915.1 - 6 - 7 471 80
AK290152.1 - 6 - 7 895 80
AK313161.1 - 6 - 7 493 80
AK315279.1 - 6 - 7 748 80
BC000148.2 - 6 - 7 481 80
BC003686.1 - 6 - 7 500 80
BC022890.1 - 6 - 7 505 80
BI918294.1 - 6 - 7 517 80
BT006916.1 - 5 - 6 427 80
CR457212.1 - 5 - 6 427 80
Y09567.1 - 5 - 6 427 80

View Full Product Details