Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C5orf45
Entrez Gene ID
51149
Gene Name
chromosome 5 open reading frame 45
Gene Aliases
-
Chromosome Location
Chr.5: 179837266 - 179858840 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017987.2 NP_001017987.1 3 - 4 319 145
NM_016175.3 NP_057259.2 5 - 6 484 145
GenBank mRNA AF153685.1 - 1 - 2 105 145
AK293901.1 - 5 - 6 450 145
AK295104.1 - 3 - 4 312 145
AK298954.1 - 3 - 4 276 145
BC056889.1 - 4 - 5 348 145
BC069051.1 - 5 - 6 452 145
BQ642639.1 - 3 - 4 275 145
BX537968.1 - 6 - 7 585 145
BX647375.1 - 5 - 6 452 145

View Full Product Details