The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017987.2 | NP_001017987.1 | 1 - 2 | 109 | 88 |
NM_016175.3 | NP_057259.2 | 1 - 2 | 109 | 88 | |
GenBank mRNA | AK293901.1 | - | 1 - 2 | 75 | 88 |
AK295104.1 | - | 1 - 2 | 102 | 88 | |
AK298954.1 | - | 1 - 2 | 66 | 88 | |
BC056889.1 | - | 1 - 2 | 62 | 88 | |
BC069051.1 | - | 1 - 2 | 77 | 88 | |
BF965313.1 | - | 1 - 2 | 110 | 88 | |
BX537968.1 | - | 1 - 2 | 130 | 88 | |
BX647375.1 | - | 1 - 2 | 77 | 88 | |
CR936785.1 | - | 1 - 2 | 130 | 88 |