Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC35B1
Entrez Gene ID
10237
Gene Name
solute carrier family 35 member B1
Gene Aliases
UGTREL1
Chromosome Location
Chr.17: 49700934 - 49708173 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005256929.3 XP_005256986.1 1 - 2 63 97
GenBank mRNA BX641166.1 - 1 - 2 95 97

View Full Product Details